missing translation for 'onlineSavingsMsg'
Learn More

Citrazinic acid, 98%

Product Code. 10266120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10266120 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10266120 Supplier Thermo Scientific Acros Supplier No. 167011000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99-11-6 | C6H5NO4 | 155.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 99-11-6
Melting Point >300°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C6H5NO4
Linear Formula (HO)2(C5H2N)COOH
MDL Number MFCD00006274
Quantity 100 g
Beilstein 22, 254
Merck Index 15, 2324
Synonym citrazinic acid, 2,6-dihydroxyisonicotinic acid, 2,6-dihydroxypyridine-4-carboxylic acid, 2,6-dihydroxy-4-carboxypyridine, 2,6-dihydroxy-4-pyridinecarboxylic acid, kyselina citrazinova, 2,6-dihydroxyisonicotinate, 4-pyridinecarboxylic acid, 2,6-dihydroxy, citrazinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-hydroxy-2-oxo
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alkali hydroxide or carbonate solns.,slightly soluble in hot hcl
InChI Key CSGQJHQYWJLPKY-UHFFFAOYSA-N
SMILES OC(=O)C1=CC(=O)NC(O)=C1
IUPAC Name 2-hydroxy-6-oxo-1H-pyridine-4-carboxylic acid
Molecular Weight (g/mol) 155.11
PubChem CID 7425
Formula Weight 155.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Citrazinic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Citrazinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.