missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Citrate Synthase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580505
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human citrate synthase/CS.
The protein encoded by this gene is a Krebs tricarboxylic acid cycle enzyme that catalyzes the synthesis of citrate from oxaloacetate and acetyl coenzyme A. The enzyme is found in nearly all cells capable of oxidative metabolism. This protein is nuclear encoded and transported into the mitochondrial matrix, where the mature form is found.
Specifications
| Citrate Synthase | |
| Polyclonal | |
| Unconjugated | |
| CS | |
| 2610511A05Rik; 9030605P22Rik; Ahl4; BB234005; Cis; citrate (Si)-synthase; citrate synthase; citrate synthase precursor (EC 4.1.3.7); citrate synthase, mitochondrial; citrate synthetase; Cs; wu:fb58e04; zgc:55507 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1431 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| O75390 | |
| CS | |
| Recombinant Human citrate synthase/CS Protein (Met1-Gly466). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction