missing translation for 'onlineSavingsMsg'
Learn More

Citrate Synthase Polyclonal Antibody
GREENER_CHOICE

Citrate Synthase Polyclonal Antibody

Brand:  Invitrogen PA576803

Product Code. 15913325

  • £481.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Citrate Synthase Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Citrate Synthase
Polyclonal
Unconjugated
CS
2610511A05Rik; 9030605P22Rik; Ahl4; BB234005; Cis; citrate (Si)-synthase; citrate synthase; citrate synthase precursor (EC 4.1.3.7); citrate synthase, mitochondrial; citrate synthetase; Cs; wu:fb58e04; zgc:55507
Rabbit
Antigen Affinity Chromatography
RUO
12974, 1431, 170587
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O75390, Q8VHF5, Q9CZU6
CS
Recombinant full length Human Citrate Synthase.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.