missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Citrate Synthase Monoclonal Antibody (CL2545)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA534991
This item is not returnable.
View return policy
Description
Immunogen sequence: ADLIPKEQAR IKTFRQQHGK TVVGQITVDM MYGGMRGMKG LVYETSVLDP DEGIRFRGFS IPECQKLLPK AKGGEEPLPE GLFWLLVTGH IPTEEQVSWL Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 94%.
The protein encoded by this gene is a Krebs tricarboxylic acid cycle enzyme that catalyzes the synthesis of citrate from oxaloacetate and acetyl coenzyme A. The enzyme is found in nearly all cells capable of oxidative metabolism. This protein is nuclear encoded and transported into the mitochondrial matrix, where the mature form is found.
Specifications
| Citrate Synthase | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75390 | |
| CS | |
| Recombinant protein fragment of Human CS (Product #RP-97646) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CL2545 | |
| Unconjugated | |
| CS | |
| 2610511A05Rik; 9030605P22Rik; Ahl4; BB234005; Cis; citrate (Si)-synthase; citrate synthase; citrate synthase precursor (EC 4.1.3.7); citrate synthase, mitochondrial; citrate synthetase; Cs; wu:fb58e04; zgc:55507 | |
| Mouse | |
| Protein A | |
| RUO | |
| 1431 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction