missing translation for 'onlineSavingsMsg'
Learn More

Citral, TRC

Product Code. 17934275
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17934275 100 g 100g
17973925 5 g 5g
17973465 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17934275 Supplier Toronto Research Chemicals Supplier No. TRCC520000100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5392-40-5
Molecular Formula C10 H16 O
Molecular Weight (g/mol) 152.23
Synonym 3,7-Dimethyl-2,6-octadienal, 3,7-Dimethyl-2,6-octadien-1-al, Citral, Citral PQ Extra, Lemarome N, Lemsyn GB, NSC 6170
IUPAC Name 3,7-dimethylocta-2,6-dienal
SMILES CC(=CCCC(=CC=O)C)C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H16O/c1-9(2)5-4-6-10(3)7-8-11/h5,7-8H,4,6H2,1-3H3
Formula Weight 152.1201
Percent Purity >95%
Chemical Name or Material Citral
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.