missing translation for 'onlineSavingsMsg'
Learn More

Citraconic anhydride, 98%

Product Code. 11367355
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367355 500 g 500g
11341857 25 g 25g
11357355 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367355 Supplier Thermo Scientific Alfa Aesar Supplier No. L05238.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 616-02-4 | C5H4O3 | 112.084 g/mol

Citraconic anhydride is used as a monomer for specialty unsaturated polyester resins. It is used to prepare bicyclic pyrrolidines, maleimides and co- and terpolymers. It is also used for the protection of N-terminal amino acids. Further, it is involved in the preparation of clothing ping acid and its amide. In addition to this, it is selectively used for the isolation of histidyl peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citraconic anhydride is used as a monomer for specialty unsaturated polyester resins. It is used to prepare bicyclic pyrrolidines, maleimides and co- and terpolymers. It is also used for the protection of N-terminal amino acids. Further, it is involved in the preparation of clothing ping acid and its amide. In addition to this, it is selectively used for the isolation of histidyl peptides.

Solubility
Miscible with water, alcohol, acetone and ether.

Notes
Moisture sensitive. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-02-4
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.084
MDL Number MFCD00005522
InChI Key AYKYXWQEBUNJCN-UHFFFAOYSA-N
Synonym citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx
PubChem CID 12012
IUPAC Name 3-methylfuran-2,5-dione
SMILES CC1=CC(=O)OC1=O

Specifications

Melting Point 7°C to 8°C
Density 1.243
Boiling Point 213°C to 214°C
Flash Point 101°C (213°F)
Refractive Index 1.471
Quantity 500 g
UN Number UN2810
Beilstein 1835
Sensitivity Moisture sensitive
Solubility Information Miscible with water,alcohol,acetone and ether.
Formula Weight 112.08
Percent Purity 98%
Chemical Name or Material Citraconic anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.