missing translation for 'onlineSavingsMsg'
Learn More

Citraconic Acid, TRC

Product Code. 30355404
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.50g
250mg
500mg
Product Code. Quantity unitSize
30355404 2.5 g 2.50g
30353447 250 mg 250mg
30363693 500 mg 500mg
3 options
This item is not returnable. View return policy

Product Code. 30355404

Brand: Toronto Research Chemicals TRCC5205182.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-23-7
Formula Weight 130.0266
InChI Formula InChI=1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h2H,1H3,(H,6,7)(H,8,9)/b3-2-
IUPAC Name (Z)-2-methylbut-2-enedioic acid
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.09
SMILES C\C(=C\C(=O)O)\C(=O)O
Synonym (2Z)-2-Methyl-2-butenedioic Acid, (Z)-2-Methyl-2-butenedioic Acid, 2-Methylmaleic Acid, Methylmaleic Acid
Chemical Name or Material Citraconic Acid

Specifications

Quantity 2.5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.