missing translation for 'onlineSavingsMsg'
Learn More

Citraconic acid, MedChemExpress

Product Code. 16427507
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16427507 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16427507 Supplier Medchem Express Supplier No. HY113298
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Citraconic acid belongs to the class of organic compounds known as methyl-branched fatty acids.

Citraconic acid belongs to the class of organic compounds known as methyl-branched fatty acids.

Specifications

Chemical Name or Material Citraconic acid
CAS 498-23-7
Color White
Purity Grade Notes Research
Molecular Formula C5H6O4
Quantity 5 g
Solubility Information DMSO : ≥ 100 mg/mL (768.64 mM)
SMILES O=C(O)/C(C)=C\C(O)=O
Molecular Weight (g/mol) 130.1
Formula Weight 130.1
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.