missing translation for 'onlineSavingsMsg'
Learn More

Citiolone, MedChemExpress

Product Code. 16408526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
16408526 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 16408526

Brand: Medchem Express HYB0931

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Citiolone is a derivative of the amino acid cysteine, used in liver therapy.

Citiolone is a derivative of the amino acid cysteine, used in liver therapy.

Specifications

Chemical Name or Material Citiolone
Physical Form Solid
CAS 1195-16-0
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H9NO2S
Solubility Information DMSO : ≥ 50 mg/mL (314.05 mM)
SMILES CC(NC(CCS1)C1=O)=O
Molecular Weight (g/mol) 159.21
Formula Weight 159.21
Percent Purity 98.36%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.