missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citidine Deaminase Antibody (S08-4C9), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | Citidine Deaminase |
|---|---|
| Clone | S08-4C9 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:500, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650695
|
Novus Biologicals
NBP3-19948-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696834
|
Novus Biologicals
NBP3-19948-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Citidine Deaminase Monoclonal antibody specifically detects Citidine Deaminase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Citidine Deaminase | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 978 | |
| IgG | |
| Affinity purified |
| S08-4C9 | |
| Western Blot, Immunohistochemistry 1:500, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CDDCytidine aminohydrolase, cytidine deaminase, cytosine nucleoside deaminase, EC 3.5.4.5, small cytidine deaminase | |
| Recombinant protein of human Citidine Deaminase (Uniprot # P32320) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title