missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citidine Deaminase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£143.00 - £251.00
Specifications
| Antigen | Citidine Deaminase |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:10000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633807
|
Novus Biologicals
NBP3-12595-100ul |
100 μg |
£251.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688498
|
Novus Biologicals
NBP3-12595-50ul |
50 μg |
£143.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Citidine Deaminase Polyclonal antibody specifically detects Citidine Deaminase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Citidine Deaminase | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, pH7.0 | |
| 978 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CDDCytidine aminohydrolase, cytidine deaminase, cytosine nucleoside deaminase, EC 3.5.4.5, small cytidine deaminase | |
| Produced in rabbits immunized with E. coli-derived Human Citidine Deaminase fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title