missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CITED4 Monoclonal Antibody (HT13-2D6.3), DyLight™ 650

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516075
This item is not returnable.
View return policy
Description
Suggested positive control: ECV304 cell lysate, transfected whole cell Hep3B lysate.
Cited4 is a member of The CITED family proteins that bind to CBP/p300 transcriptional integrators through their conserved C-terminal acidic domain and function as coactivators. It also interacts with isoforms of the TFAP2 transcription factor, coactivating TFAP2-dependent transcription.
Specifications
| CITED4 | |
| Monoclonal | |
| 1 mg/mL | |
| 50mM sodium borate with 0.05% sodium azide | |
| Q96RK1 | |
| CITED4 | |
| N-terminal fusion protein to full-length human CITED4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| HT13-2D6.3 | |
| DyLight 650 | |
| CITED4 | |
| Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4; cbp/p300-interacting transactivator 4; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 4; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4; cited4; cited4 {ECO:0000312; Mrg2; MRG-2; MSG1-related protein 2; RGD:620113}; transcriptional co-activator 4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 163732 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction