missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED4 Antibody (HT13-2D6.3), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-41572G
This item is not returnable.
View return policy
Description
CITED4 Monoclonal specifically detects CITED4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CITED4 | |
| Monoclonal | |
| DyLight 488 | |
| Q96RK1 | |
| CITED4 | |
| N-terminal fusion protein to full-length human CITED4 [UniProt# Q96RK1] | |
| 0.1 mL | |
| Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators | |
| 163732 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HT13-2D6.3 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| cbp/p300-interacting transactivator 4, Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain4,MRG-2, MRG2, MSG1-related protein 2, transcriptional co-activator 4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction