missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CITED2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585562
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (99%), Rat (100%), Sheep (98%), Bovine (98%). Positive Control: 293T, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene inhibits transactivation of HIF1A-induced genes by competing with binding of HIF1a to p300-CH1. Mutations in this gene are a cause of cardiac septal defects. Three transcript variants encoding the same protein have been found for this gene.
Specifications
| CITED2 | |
| Polyclonal | |
| Unconjugated | |
| CITED2 | |
| AI835299; ASD8; Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2; cbp/p300-interacting transactivator 2; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 2; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2; Cited2; ER154-like; melanocyte specific gene 2; melanocyte-specific gene 1-related gene 1; Mrg1; MRG-1; MSG1-related gene 1; Msg2; MSG-related protein 1; P35srj; VSD2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10370 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q99967 | |
| CITED2 | |
| Recombinant protein encompassing a sequence within the center region of human CITED2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction