missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26999-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.Specifications
| CITED1 | |
| Polyclonal | |
| Unconjugated | |
| CITED1 | |
| CITED1, Melanocyte specific protein 1, MSG1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4435, 64466 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99966 | |
| CITED1 | |
| CITED1 Fusion Protein Ag24643 | |
| 20 μL | |
| Primary | |
| Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction