Learn More
CITED1 Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL555-26999-100UL
Description
The CITED1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CITED1. This antibody recognizes human, rat, mouse antigen. The CITED1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™555.
Specifications
| CITED1 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |
| Immunofluorescence | |
| CoraLite 555 | |
| 100 μL | |
| 4435 | |
| IgG |