missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CITED1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540585
This item is not returnable.
View return policy
Description
Peptide sequence: IGSPTTTPPT KPPSFNLHPA PHLLASMQLQ KLNSQYQGMA AATPGQPGEA Sequence homology: Cow: 85%; Dog: 85%; Guinea Pig: 85%; Human: 100%; Mouse: 77%; Rabbit: 83%; Rat: 85%.
This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.
Specifications
| CITED1 | |
| Polyclonal | |
| Unconjugated | |
| CITED1 | |
| AI316840; AU019144; Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1; cbp/p300-interacting transactivator 1; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 1; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1; Cited1; CITED1 protein; melanocyte-specific gene 1; melanocyte-specific gene 1 protein; melanocyte-specific protein 1; melanocyte-specific protein 1; p35srj; CBP/P300-interacting transactivator 1; CITED1; Msg1; MSG1 protein; transactivator, with Glu/Asp-rich carboxy-terminal domain, 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4435 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99966 | |
| CITED1 | |
| Synthetic peptide directed towards the middle region of human CITED1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction