missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CITED1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565541
This item is not returnable.
View return policy
Description
Immunogen sequence: MPTTSRPALDV KGGTSPAKED ANQEMSSVAY SNLAVKDRKA VAILHYPGVA SNGTKASGAP TSSSGSPIGS PTTTPPTKP Highest antigen sequence identity to the following orthologs - mouse 77%, rat 76%.
This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.
Specifications
| CITED1 | |
| Polyclonal | |
| Unconjugated | |
| CITED1 | |
| AI316840; AU019144; Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1; cbp/p300-interacting transactivator 1; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 1; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1; Cited1; CITED1 protein; melanocyte-specific gene 1; melanocyte-specific gene 1 protein; melanocyte-specific protein 1; melanocyte-specific protein 1; p35srj; CBP/P300-interacting transactivator 1; CITED1; Msg1; MSG1 protein; transactivator, with Glu/Asp-rich carboxy-terminal domain, 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4435 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99966 | |
| CITED1 | |
| Recombinant Human CITED1. Recombinant protein control fragment (Product #RP-106216). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction