missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED1, Mouse anti-Human, Clone: 6G8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004435-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: SMQLQKLNSQYQGMAAATPGQPGEAGPLQNWDFGAQAGGAESLSPSAGAQSPAIIDSDPVDEEVLMSLVVELGLDRANELPELWLGQNEFDFTADFPSSCSpecifications
| CITED1 | |
| Monoclonal | |
| Unconjugated | |
| Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1 | |
| MSG1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, KnockDown, Western Blot | |
| 6G8 | |
| In 1x PBS, pH 7.4 | |
| NM_004143 | |
| CITED1 | |
| CITED1 (NP_004134, 94 a.a. ∼ 193 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4435 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction