missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CITCO

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CITCO is a constitutive androstane receptor (CAR) agonist (EC50 = 49 nM). Displays > 100-fold selectivity over PXR receptors and no activity at LXR, ERα, ERβ, PPAR, RAR, FXR, VDR and THR. Induces CAR nuclear translocation and expression of CYP2B6 in hepatocytes in vitro.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | CITCO |
| Purity | >98% |
| Molecular Weight (g/mol) | 436.74 |
| Inhibitors | CITCO |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction