missing translation for 'onlineSavingsMsg'
Learn More

Citarinostat, MedChemExpress

Product Code. 16476958
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476958 10 mg 10mg
16443017 50 mg 50mg
16429507 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16476958 Supplier Medchem Express Supplier No. HY15994
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Citarinostat (ACY241) is a second generation potent, orally active and high-selective HDAC6 inhibitor with an IC50 of 2.6 nM (IC50s of 35 nM, 45 nM, 46 nM and 137 nM for HDAC1, HDAC2, HDAC3 and HDAC8, respectively). Citarinostat has anticancer effects.

Citarinostat (ACY241) is a second generation potent, orally active and high-selective HDAC6 inhibitor with an IC50 of 2.6 nM (IC50s of 35 nM, 45 nM, 46 nM and 137 nM for HDAC1, HDAC2, HDAC3 and HDAC8, respectively). Citarinostat has anticancer effects.

Specifications

Chemical Name or Material Citarinostat
CAS 1316215-12-9
Color White
Purity Grade Notes Research
Molecular Formula C24H26ClN5O3
Quantity 10 mg
Synonym ACY241
Solubility Information DMSO : ≥ 30 mg/mL (64.11 mM)
SMILES O=C(C1=CN=C(N(C2=CC=CC=C2Cl)C3=CC=CC=C3)N=C1)NCCCCCCC(NO)=O
Molecular Weight (g/mol) 467.95
Formula Weight 467.95
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.