missing translation for 'onlineSavingsMsg'
Learn More

Citalopram hydrobromide, MedChemExpress

Product Code. 16488716
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488716 100 mg 100mg
16478688 10 mg 10mg
16444577 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488716 Supplier Medchem Express Supplier No. HYB1287
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Citalopram hydrobromide is a selective serotonin reuptake inhibitor (SSRI). Citalopram hydrobromide inhibits 5-HT uptake into synaptosomes with an IC50 of 1.8 nM. Citalopram hydrobromideinhibits the 5-HT uptake in rabbit blood platelets with an IC50 of 14 nM. Antidepressant effect.

Citalopram hydrobromide is a selective serotonin reuptake inhibitor (SSRI). Citalopram hydrobromide inhibits 5-HT uptake into synaptosomes with an IC50 of 1.8 nM. Citalopram hydrobromideinhibits the 5-HT uptake in rabbit blood platelets with an IC50 of 14 nM. Antidepressant effect.

Specifications

Chemical Name or Material Citalopram hydrobromide
CAS 59729-32-7
Color White
Purity Grade Notes Research
Molecular Formula C20H22BrFN2O
Quantity 100 mg
Synonym (±)-Citalopram hydrobromide Lu 10-171
Solubility Information DMSO : ≥ 38 mg/mL (93.76 mM) ∣H2O : 10 mg/mL (24.67 mM; Need ultrasonic)
SMILES N#CC1=CC2=C(C(C3=CC=C(F)C=C3)(CCCN(C)C)OC2)C=C1.[H]Br
Molecular Weight (g/mol) 405.3
Formula Weight 405.3
Percent Purity 99.66%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.