missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cisplatin, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Cisplatin (CDDP) is an antineoplastic chemotherapy agent by cross-linking with DNA and causing DNA damage in cancer cells. Cisplatin activates ferroptosis and induces autophagy.
Specifications
Specifications
| Chemical Name or Material | Cisplatin |
| Physical Form | Powder |
| CAS | 15663-27-1 |
| Color | Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | Cl2H6N2Pt |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | cis-Platinum CDDP cis-Diaminodichloroplatinum |
| Solubility Information | DMSO : 250 mg/mL (833.19 mM; Need ultrasonic; DMSO can inactivate Cisplatin's activity) ∣DMF : 5 mg/mL (16.66 mM; Need ultrasonic; DMSO can inactivate Cisplatin's activity) ∣H2O : 1 mg/mL (3.33 mM; Need ultrasonic; DMSO can inactivate Cisplatin's activity) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction