missing translation for 'onlineSavingsMsg'
Learn More

CISH/CIS-1 Antibody (989616) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30731931 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731931 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731931 Supplier Novus Biologicals Supplier No. FAB3194MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CISH/CIS-1 Monoclonal antibody specifically detects CISH/CIS-1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CISH/CIS-1
Applications Western Blot
Classification Monoclonal
Clone 989616
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias CIS-1cytokine-inducible SH2-containing protein, CIScytokine-inducible inhibitor of signaling type 1B, cytokine inducible SH2-containing protein, G18Protein G18, SOCS, Suppressor of cytokine signaling
Host Species Rat
Immunogen E.coli-derived recombinant human CIS-1, Leu11-Leu258, Accession # Q9NSE2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1154
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.