Learn More
CISD3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30480-1-AP-20UL
This item is not returnable.
View return policy
Description
The CISD3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CISD3. This antibody recognizes human, mouse antigen. The CISD3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CISD3 | |
| 450 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| CDGSH iron sulfur domain 3, CISD3, Miner2, MitoNEET related protein 2 | |
| 20 μL | |
| 284106 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |