missing translation for 'onlineSavingsMsg'
Learn More

CISD2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18427546 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427546 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427546 Supplier Novus Biologicals Supplier No. NBP281907AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CISD2 Polyclonal antibody specifically detects CISD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CISD2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias CDGSH iron sulfur domain 2, CDGSH iron-sulfur domain-containing protein 2, CDGSH2, Endoplasmic reticulum intermembrane small protein, ERISWFS2, Miner1Wolfram syndrome 2, mitoNEET related 1, MitoNEET-related 1 protein, NAF-1, Nutrient-deprivation autophagy factor-1, ZCD2CDGSH iron sulfur domain-containing protein 2, zinc finger, CDGSH-type domain 2
Host Species Rabbit
Immunogen CISD2 antibody was raised against a 15 amino acid peptide near the center of human CISD2. The immunogen is located within amino acids 40 - 90 of CISD2. Amino Acid Squence: VRPFLPKKKQQKDSL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 493856
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.