missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CISD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106281
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CISD1.
This gene encodes a protein with a CDGSH iron-sulfur domain and has been shown to bind a redox-active [2Fe-2S] cluster. The encoded protein has been localized to the outer membrane of mitochondria and is thought to play a role in regulation of oxidation. Genes encoding similar proteins are located on chromosomes 4 and 17, and a pseudogene of this gene is located on chromosome 2.
Specifications
| CISD1 | |
| Polyclonal | |
| Unconjugated | |
| cisd1 | |
| AU043990; AW743335; C10orf70; CDGSH iron sulfur domain 1; CDGSH iron sulfur domain-containing protein 1; CDGSH iron sulfur domain-containing protein-like protein; CDGSH iron-sulfur domain-containing protein 1; Cisd1; D10Ertd214e; hypothetical protein LOC393577; hypothetical protein LOC531444; MDS029; MGC14684; MitoNEET; RGD1309529; si:dkey-18n13.4; ZCD1; zgc:63561; zinc finger CDGSH-type domain 1; zinc finger, CDGSH-type domain 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 294362, 393577, 52637, 55847 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B0K020, Q91WS0, Q9NZ45 | |
| cisd1 | |
| A synthesized peptide derived from human CISD1(Accession Q9NZ45), corresponding to amino acid residues K55-K105. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction