missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CISD1 Antibody (OTI4G3), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72026PCP
This item is not returnable.
View return policy
Description
CISD1 Monoclonal antibody specifically detects CISD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| CISD1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI4G3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| C10orf70, CDGSH iron sulfur domain 1, CDGSH iron sulfur domain-containing protein 1, CDGSH iron-sulfur domain-containing protein 1, chromosome 10 open reading frame 70, MDS029, MGC14684, mitoNEET, ZCD1, zinc finger CDGSH-type domain 1, zinc finger, CDGSH-type domain 1 | |
| Full length human recombinant protein of human CISD1 (NP_060934) produced in HEK293T cell. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 55847 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction