missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CISD1 Monoclonal antibody specifically detects CISD1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
Specifications
| Antigen | CISD1 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 1A8 |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C10orf70, CDGSH iron sulfur domain 1, CDGSH iron sulfur domain-containing protein 1, CDGSH iron-sulfur domain-containing protein 1, chromosome 10 open reading frame 70, MDS029, MGC14684, mitoNEET, ZCD1, zinc finger CDGSH-type domain 1, zinc finger, CDGSH-type domain 1 |
| Host Species | Mouse |
| Immunogen | Recombinant human CISD1 (32-108aa) purified from E. coli |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?