missing translation for 'onlineSavingsMsg'
Learn More

Cisapride, MedChemExpress

Product Code. 16486396
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486396 100 mg 100mg
16485968 10 mg 10mg
16452117 50 mg 50mg
16451116 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486396 Supplier Medchem Express Supplier No. HY14149

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cisapride is an orally and potent 5-HT4 receptor agonist and hERG inhibitor. Cisapride is an prokinetic agent which facilitates or restores motility throughout the length of the gastrointestinal tract. Cisapride stimulates gastrointestinal motor activity through an indirect mechanism involving the release of acetylcholine mediated by postganglionic nerve endings in the myenteric plexus of the gut.

Cisapride is an orally and potent 5-HT4 receptor agonist and hERG inhibitor. Cisapride is an prokinetic agent which facilitates or restores motility throughout the length of the gastrointestinal tract. Cisapride stimulates gastrointestinal motor activity through an indirect mechanism involving the release of acetylcholine mediated by postganglionic nerve endings in the myenteric plexus of the gut.

Specifications

Chemical Name or Material Cisapride
CAS 81098-60-4
Color White
Purity Grade Notes Research
Molecular Formula C23H29ClFN3O4
Quantity 100 mg
Synonym R 51619 (±)-Cisaprid
Solubility Information DMSO : 100 mg/mL (214.62 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.21 mM; Need ultrasonic and warming)
SMILES ClC1=C(N)C=C(OC)C(C(N[C@H]2[C@@H](OC)CN(CCCOC3=CC=C(F)C=C3)CC2)=O)=C1
Molecular Weight (g/mol) 465.95
Formula Weight 465.95
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.