missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ cis-VZ 185
GREENER_CHOICE

Product Code. 18798606
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 18798606

Brand: Tocris Bioscience™ 6939/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Negative control for VZ 185 (Cat. No. 6936)

cis-VZ 185 is a negative control for VZ 185 (Cat. No. 6936).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S,4 S)-N-(2-((5-(4-(2,6-Dimethoxy-4-(2-methyl-1-oxo-1,2-dihydro-2,7-naphthyridin-4-yl)benzyl)piperazin-1-yl)pentyl)oxy)-4-(4-methylthiazol-5-yl)benzyl)-1-(-2-(1-fluorocyclopropane-1-carboxamido)-3,3-dimethylbutanoyl)-4-hydroxypyrrolidine-2-carboxamide
CAS 2306193-98-4
Quantity 5 mg
Target Active Degrader Negative Controls
Molecular Formula C53H67FN8O8S
Formula Weight 2576
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.