missing translation for 'onlineSavingsMsg'
Learn More

cis/trans-Nepetalactol, TRC

Product Code. 30482593
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482593 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482593 Supplier Toronto Research Chemicals Supplier No. TRCN390065100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4aS,7S,7aR)-Nepetalactol
CAS 109215-55-6
Molecular Formula C10 H16 O2
Quantity 100 mg
Synonym Cyclopenta[c]pyran-1-ol','1,4a,5,6,7,7a-hexahydro-4,7-dimethyl-','(4aS,7S,7aR)-, Cyclopenta[c]pyran-1-ol','1,4a,5,6,7,7a-hexahydro-4,7-dimethyl- (9CI), (4aS,7S,7aR)-1,4a,5,6,7,7a-Hexahydro-4,7-dimethylcyclopenta[c]pyran-1-ol, (4aS,7S,7aR)-Nepetalactol, Nepetalactol, cis/trans-Nepetalactol
InChI Formula InChI=1S/C10H16O2/c1-6-3-4-8-7(2)5-12-10(11)9(6)8/h5-6,8-11H,3-4H2,1-2H3/t6-,8+,9+,10?/m0/s1
SMILES C[C@H]1CC[C@H]2[C@@H]1C(O)OC=C2C
IUPAC Name (4aS,7S,7aR)-4,7-dimethyl-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-1-ol
Molecular Weight (g/mol) 168.23
Formula Weight 168.115
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.