missing translation for 'onlineSavingsMsg'
Learn More

(+)-cis,trans-Abscisic Acid, 98%, TRC

Product Code. 17944045
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944045 5 mg 5mg
17911485 10 mg 10mg
17963515 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17944045 Supplier Toronto Research Chemicals Supplier No. TRCA1100005MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21293-29-8
Molecular Formula C15H20O4
Molecular Weight (g/mol) 264.32
Synonym 2,4-Pentadienoic acid, 5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl]-3-methyl-, (2Z,4E)-, 2,4-Pentadienoic acid, 5-(1-hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-, (Z,E)-(S)-(+)- (8CI), 2,4-Pentadienoic acid, 5-(1-hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-, [S-(Z,E)]-, (2Z,4E)-5-[(1S)-1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl]-3-methyl-2,4-pentadienoic acid, (+)-(S)-Abscisic acid, (+)-(cis,trans)-Abscisic acid, (+)-ABA, (+)-Abscisic acid, (+)-Abscisin II, (+)-cis-Abscisic acid, (S)-(+)-ABA, (S)-(+)-Abscisic acid, (S)-ABA, (Z,E)-Abscisic acid, 2-cis,4-trans-Abscisic acid, ABA, ABK, Abscisic acid, Abscisic acid, (+)-, Abscisin II, Dormin, Dormin (abscission factor), NSC 146877, NSC 148832, PBI 58, ProTone, cis,trans-Abscisic acid, cis-Abscisic acid, cis-trans-(+)-Abscisic acid, (+)-cis,trans-Abscisic acid
IUPAC Name (2Z,4E)-5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxo-cyclohex-2-en-1-yl]-3-methyl-penta-2,4-dienoic acid
SMILES C\C(=C\C(=O)O)\C=C\[C@@]1(O)C(=CC(=O)CC1(C)C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H20O4/c1-10(7-13(17)18)5-6-15(19)11(2)8-12(16)9-14(15,3)4/h5-8,19H,9H2,1-4H3,(H,17,18)/b6-5+,10-7-/t15-/m1/s1
Formula Weight 264.1362
Percent Purity >95%
Chemical Name or Material (+)-Abscisic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.