missing translation for 'onlineSavingsMsg'
Learn More

cis,trans-(1,1-Dimethylethoxy)carbonyl Tranexamic Acid-13C2,15N, TRC

Product Code. 30387322
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387322 10 mg
30381733 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387322 Supplier Toronto Research Chemicals Supplier No. TRCD47300710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346604-84-9
Molecular Formula C1113C2H2315NO4
Molecular Weight (g/mol) 260.3
Synonym 4-[[[(1,1-Dimethylethoxy)carbonyl]amino]methyl]-cyclohexanecarboxylic Acid-13C2,15N, 4-[[(tert-Butoxycarbonyl)amino]methyl]cyclohexanecarboxylic Acid-13C2,15N
SMILES O[13C](C1CCC([13CH2][15NH]C(OC(C)(C)C)=O)CC1)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H23NO4/c1-13(2,3)18-12(17)14-8-9-4-6-10(7-5-9)11(15)16/h9-10H,4-8H2,1-3H3,(H,14,17)(H,15,16)/i8+1,11+1,14+1
Chemical Name or Material cis,trans-(1,1-Dimethylethoxy)carbonyl Tranexamic Acid-13C2,15N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.