missing translation for 'onlineSavingsMsg'
Learn More

cis-Tetrahydrophthalimide, TRC

Product Code. 30416644
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416644 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416644 Supplier Toronto Research Chemicals Supplier No. TRCT29424010G
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-1,2,3,6-Tetrahydrophthalimide
CAS 1469-48-3
Molecular Formula C8 H9 N O2
Quantity 10 g
Synonym 1H-Isoindole-1,3(2H)-dione','3a,4,7,7a-tetrahydro-','(3aR,7aS)-rel-, 1H-Isoindole-1,3(2H)-dione','3a,4,7,7a-tetrahydro-','cis-, 4-Cyclohexene-1,2-dicarboximide','cis- (8CI), rel-(3aR,7aS)-3a,4,7,7a-Tetrahydro-1H-isoindole-1,3(2H)-dione, NSC 41605, cis-1,2,3,6-Tetrahydrophthalimide, cis-4-Cyclohexene-1,2-dicarboximide, cis-Tetrahydrophthalimide, cis-Δ4-Tetrahydrophthalimide
InChI Formula InChI=1S/C8H9NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-2,5-6H,3-4H2,(H,9,10,11)/t5-,6+
SMILES O=C1NC(=O)[C@H]2CC=CC[C@@H]12
IUPAC Name (3aR,7aS)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
Molecular Weight (g/mol) 151.16
Formula Weight 151.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.