missing translation for 'onlineSavingsMsg'
Learn More

cis-Nonachlor, SPEX CertiPrep™

Product Code. 15236216 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 15236216

Brand: SPEX Certiprep S2765

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
cis-Nonachlor5103-73-11000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 5103-73-1, 67-56-1
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Packaging 2mL amber prescored glass ampule
Molecular Formula C10H5Cl9
Quantity 1 mL
Synonym cis-nonachlor, cis-nonachlor solution, cis-nonachlor standard, trans-nonachlor solution, trans-nonachlor standard, cis-nonachlor 10 microg/ml in cyclohexane, cis-nonachlor 100 microg/ml in isooctane, trans-nonachlor 10 microg/ml in cyclohexane, trans-nonachlor 100 microg/ml in cyclohexane, cis-nonachlor solution, 100 mug/ml in hexane, analytical standard
InChI Key OCHOKXCPKDPNQU-MSFPVELZSA-N
SMILES C12C(C(C(C1Cl)Cl)Cl)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl
Molecular Weight (g/mol) 444.2
PubChem CID 12313426
Chemical Name or Material cis-Nonachlor
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • cis-Nonachlor
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.