missing translation for 'onlineSavingsMsg'
Learn More

cis-N-Boc-4-cyano-L-proline methyl ester, 97%

Product Code. 15419527
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419527 250 mg 250mg
15409527 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419527 Supplier Thermo Scientific Alfa Aesar Supplier No. H52783.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 487048-28-2 | C12H18N2O4 | 254.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487048-28-2
Molecular Formula C12H18N2O4
Molecular Weight (g/mol) 254.29
MDL Number MFCD03787928
InChI Key GHLKJIQVORAVHE-BDAKNGLRSA-N
Synonym n-boc-cis-4-cyano-l-proline methyl ester, 2s,4s-1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, boc-cis-4-cyano-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4s-4-cyanopyrrolidine-1,2-dicarboxylate, 1,2-pyrrolidinedicarboxylic acid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4s, 1,2-pyrrolidinedicarboxylicacid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4s, cis-n-boc-4-cyano-l-proline methyl ester 250mg, 2s,4s-1-tert-butoxycarbonyl-4-cyano-2-methylpyrrolidine-2-carboxylic acid, 2s,4s-4-cyano-1,2-pyrrolidinedicarboxylic acid 1-tert-butyl 2-methyl ester, 1,2-pyrrolidinedicarboxylic acid,4-cyano-,1-1,1-dimethylethyl 2-methyl ester, 2s,4s
PubChem CID 17750454
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate
SMILES COC(=O)[C@@H]1C[C@@H](CN1C(=O)OC(C)(C)C)C#N

Specifications

Quantity 250 mg
Formula Weight 254.29
Percent Purity 97%
Chemical Name or Material cis-N-Boc-4-cyano-L-proline methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.