missing translation for 'onlineSavingsMsg'
Learn More

cis-Hydroxy Solifenacin (Mixture of Diastereomers), TRC

Product Code. 30397793
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397793 1 mg 1mg
30392450 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397793 Supplier Toronto Research Chemicals Supplier No. TRCH9535251MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-hydroxy Solifenacin (mixture of diastereomers)
Analyte or Component Names cis-hydroxy Solifenacin (mixture of diastereomers)
Shipping Condition Room Temperature
Molecular Formula C23 H26 N2 O3
Synonym rel-(1S,4R)-3,4-Dihydro-4-hydroxy-1-phenyl-2(1H)-isoquinolinecarboxylic Acid (3R)-1-Azabicyclo[2.2.2]oct-3-yl Ester
InChI Formula InChI=1S/C23H26N2O3/c26-20-15-25(23(27)28-21-14-16-10-12-24(21)13-11-16)22(17-6-2-1-3-7-17)19-9-5-4-8-18(19)20/h1-9,16,20-22,26H,10-15H2/t20-,21+,22-/m0/s1
Solution Type Neat
SMILES O[C@H]1CN([C@@H](c2ccccc2)c3ccccc13)C(=O)O[C@@H]4CC5CCN4CC5
IUPAC Name [(2R)-quinuclidin-2-yl] (rel-1S,4R)-4-hydroxy-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate
Industry Type Forensics
Molecular Weight (g/mol) 378.46
Quantity 1 mg
Formula Weight 378.1943
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.