missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-Ethyl Cinnamate (contains up to 10% Ethyl dihydrocinnamate), TRC
Specifications
Specifications
| Chemical Name or Material | cis-Ethyl Cinnamate |
| CAS | 4610-69-9 |
| Molecular Formula | C11 H12 O2 |
| Quantity | 50 mg |
| Synonym | 2-Propenoic acid, 3-phenyl-, ethyl ester, (2Z)-, 2-Propenoic acid, 3-phenyl-, ethyl ester, (Z)-, Cinnamic acid, ethyl ester, (Z)- (8CI), (2Z)-3-Phenylprop-2-enoic acid ethyl ester, (2Z)-3-Phenyl-2-propenoic acid ethyl ester, (Z)-3-Phenyl-2-propenoic acid ethyl ester, (Z)-Ethyl cinnamate, Ethyl (Z)-3-phenyl-2-propenoate, Ethyl (Z)-3-phenylacrylate, Ethyl (Z)-3-phenylacrylate, Ethyl (Z)-cinnamate, Ethyl allocinnamate, Ethyl cis-cinnamate, cis-3-Phenyl-2-propenoic acid ethyl ester |
| InChI Formula | InChI=1S/C11H12O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-9H,2H2,1H3/b9-8- |
| SMILES | CCOC(=O)\C=C/c1ccccc1 |
| IUPAC Name | ethyl (Z)-3-phenylprop-2-enoate |
| Molecular Weight (g/mol) | 176.21 |
| Formula Weight | 176.0837 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?