missing translation for 'onlineSavingsMsg'
Learn More

cis-Ethyl Cinnamate (contains up to 10% Ethyl dihydrocinnamate), TRC

Product Code. 30388351
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388351 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388351 Supplier Toronto Research Chemicals Supplier No. TRCE903950500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-Ethyl Cinnamate
CAS 4610-69-9
Molecular Formula C11 H12 O2
Quantity 500 mg
Synonym 2-Propenoic acid, 3-phenyl-, ethyl ester, (2Z)-, 2-Propenoic acid, 3-phenyl-, ethyl ester, (Z)-, Cinnamic acid, ethyl ester, (Z)- (8CI), (2Z)-3-Phenylprop-2-enoic acid ethyl ester, (2Z)-3-Phenyl-2-propenoic acid ethyl ester, (Z)-3-Phenyl-2-propenoic acid ethyl ester, (Z)-Ethyl cinnamate, Ethyl (Z)-3-phenyl-2-propenoate, Ethyl (Z)-3-phenylacrylate, Ethyl (Z)-3-phenylacrylate, Ethyl (Z)-cinnamate, Ethyl allocinnamate, Ethyl cis-cinnamate, cis-3-Phenyl-2-propenoic acid ethyl ester
InChI Formula InChI=1S/C11H12O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-9H,2H2,1H3/b9-8-
SMILES CCOC(=O)\C=C/c1ccccc1
IUPAC Name ethyl (Z)-3-phenylprop-2-enoate
Molecular Weight (g/mol) 176.21
Formula Weight 176.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.