missing translation for 'onlineSavingsMsg'
Learn More

(+)-cis-Diltiazem hydrochloride, Thermo Scientific™

An L-type calcium channel blocker

Brand:  Thermo Scientific Alfa Aesar J63245.09

Quantity 10g

 View more versions of this product

Product Code. 15475509

  • £128.00 / 10g
 Restricted Item
Estimated Shipment: 28-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

33286-22-5
450.978
HDRXZJPWHTXQRI-BHDTVMLSSA-N
62920
[(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride
C22H27ClN2O4S
MFCD00069252
diltiazem hydrochloride, diltiazem hcl, herbesser, dilzene, lacerol, masdil, tildiem, tiazac, mono-tildiem, bi-tildiem
CHEBI:645509
CC(=O)OC1C(SC2=CC=CC=C2N(C1=O)CCN(C)C)C3=CC=C(C=C3)OC.Cl

Specifications

33286-22-5
MFCD00069252
4228706
diltiazem hydrochloride, diltiazem hcl, herbesser, dilzene, lacerol, masdil, tildiem, tiazac, mono-tildiem, bi-tildiem
HDRXZJPWHTXQRI-BHDTVMLSSA-N
[(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride
62920
450.98
C22H27ClN2O4S
10g
14,3202
Soluble in water at 50mg/ml
CC(=O)OC1C(SC2=CC=CC=C2N(C1=O)CCN(C)C)C3=CC=C(C=C3)OC.Cl
450.978
CHEBI:645509
(+)-cis-Diltiazem hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.