missing translation for 'onlineSavingsMsg'
Learn More

cis-Dicyclohexano-18-crown-6, 98%, mixture of syn-cis and anti-cis isomers

Product Code. 10420721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10420721 2.5 g Glass bottle 2.5g
10043500 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10420721 Supplier Thermo Scientific Acros Supplier No. 277520025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16069-36-6 | C20H36O6 | 372.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16069-36-6
Molecular Formula C20H36O6
Molecular Weight (g/mol) 372.5
MDL Number MFCD00005099
InChI Key BBGKDYHZQOSNMU-UHFFFAOYSA-N
Synonym dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
PubChem CID 85955
SMILES C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2

Specifications

Melting Point 47°C to 53°C
Color White to Beige
Boiling Point 181°C to 183°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Fieser 04,142; 05,207; 06,174; 09,126; 12,147; 01,1046; 03,261; 04,442; 06,529; 12,441; 16,307
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform and acetone
Formula Weight 372.5
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material cis-Dicyclohexano-18-crown-6, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.