missing translation for 'onlineSavingsMsg'
Learn More

cis-Dichlorobis(triphenylphosphine)platinum(II), Pt 24.2% min

Product Code. 11365988
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365988 250 mg 250mg
11375988 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365988 Supplier Thermo Scientific Alfa Aesar Supplier No. 010492.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

CAS: 15604-36-1 | C36H30Cl2P2Pt | 790.57 g/mol

cis-Dichlorobis(triphenylphosphine)platinum(II) is used primarily as a reagent for the synthesis of other platinum molecules and in platinum plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15604-36-1
Molecular Formula C36H30Cl2P2Pt
Molecular Weight (g/mol) 790.57
MDL Number MFCD00010825
InChI Key ILIBUWCDNDIUOT-UHFFFAOYSA-L
Synonym cis-?dichlorobis triphenylphosphine ?platinum ii
PubChem CID 131667321
IUPAC Name platinum(2+);triphenylphosphane;dihydrochloride
SMILES Cl[Pt++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 310°C (decomposition)
Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Slightly soluble in chloroform, hexane toluene
Formula Weight 790.58
Concentration or Composition (by Analyte or Components) Pt 24.2% min.
Physical Form Crystalline
Chemical Name or Material cis-Dichlorobis(triphenylphosphine)platinum(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.