missing translation for 'onlineSavingsMsg'
Learn More

cis-Dichlorobis(diethylsulfide)platinum(II), Pt 43.0% min

Product Code. 11395978
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395978 0.5 g 0.5g
11305988 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395978 Supplier Thermo Scientific Alfa Aesar Supplier No. 010482.02
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrogenation | CAS: 15442-57-6 | C8H20Cl2PtS2 | 446.35 g/mol

Hydrogenation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15442-57-6
Molecular Formula C8H20Cl2PtS2
Molecular Weight (g/mol) 446.35
MDL Number MFCD00050943
InChI Key RFSLRYGKHBDFGC-UHFFFAOYSA-L
Synonym cis-dichlorobis diethylsulfide platinum ii, cis-dichlorobis diethyl sulfide platinum ii, dichloroplatinum, ethylsulfanylethane, trans-dichlorobis diethylsulfide platinum ii, cis-bis diethyl sulfide platinum ii chloride, dichlorobis 1,1'-thiobis ethane platinum, platinum, dichlorobis 1,1'-thiobis ethane, dichloroplatinum; ethylsulfanylethane, ethylsulfanyl ethane-dichloroplatinum 2:1
PubChem CID 167183
SMILES Cl[Pt++]Cl.CCSCC.CCSCC

Specifications

Quantity 0.5 g
Solubility Information Soluble in acetone, alcohol and benzene
IUPAC Name dichloroplatinum;ethylsulfanylethane
Formula Weight 446.37
Concentration or Composition (by Analyte or Components) Pt 43.7%
Physical Form Crystalline
Chemical Name or Material cis-Dichlorobis(diethylsulfide)platinum(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.