missing translation for 'onlineSavingsMsg'
Learn More

cis-Diamminedichloroplatinum(II), Pt 64.5% min

Product Code. 11335978
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335978 1 g 1g
11325978 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335978 Supplier Thermo Scientific Alfa Aesar Supplier No. 010471.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Potent Anticancer Agent That Blocks DNA Synthesis. | CAS: 15663-27-1 | Cl2H6N2Pt | 300.04 g/mol

Being a potent anticancer agent that blocks DNA synthesis, the pharmaceutical compositions are used in chemotherapy. First member of platinum-containing anti-cancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15663-27-1
Molecular Formula Cl2H6N2Pt
Molecular Weight (g/mol) 300.04
MDL Number MFCD00011623
InChI Key BSJGASKRWFKGMV-UHFFFAOYSA-L
Synonym Cisplatin
IUPAC Name dichloroplatinumbis(ylium) diamine
SMILES N.N.Cl[Pt++]Cl

Specifications

Melting Point 270°C (decomposition)
Color Yellow
Physical Form Micro-crystals
Quantity 1 g
Solubility Information Soluble in DMF. Insoluble in most common solvents
Formula Weight 300.06
Odor Odorless
Appearance Yellow micro crystals
Chemical Name or Material cis-Diamminedichloroplatinum(II), Potent anticancer agent that blocks DNA synthesis

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.