missing translation for 'onlineSavingsMsg'
Learn More

cis-Bis(2,2'-bipyridine)dichlororuthenium(II) dihydrate, Ru 19% min

Product Code. 11347547
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347547 5 g 5g
11357547 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347547 Supplier Thermo Scientific Alfa Aesar Supplier No. 012778.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 609844-11-3 | C20H20Cl2N4O2Ru | 520.376 g/mol

cis-Bis(2,2'-bipyridine)dichlororuthenium(II) dihydrate is used in the preparation of [Ru(m-) 2,2'-bipyridine)3]2+(PF6-)2. It is also involved in the preparation of an intermediate for the development of nanocomposite junctions with gold nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609844-11-3
Molecular Formula C20H20Cl2N4O2Ru
Molecular Weight (g/mol) 520.376
MDL Number MFCD00012040
InChI Key NHKTUSUPCAKVHT-UHFFFAOYSA-L
Synonym cis-dichlorobis 2,2'-bipyridine ruthenium ii dihydrate, 99, cis-dichlorobis 2,2'-bipyridine-ruthenium ii dihydrate
PubChem CID 71463877
IUPAC Name 2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;dihydrate
SMILES C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.O.O.[Cl-].[Cl-].[Ru+2]

Specifications

Color Black
Assay Percent Range Ru 19% min.
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 520.38 (484.35 Anhydrous)
Physical Form Crystalline
Chemical Name or Material cis-Bis-(2, 2 ft.-bipyridine)dichlororuthenium(II) dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.