missing translation for 'onlineSavingsMsg'
Learn More

cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%

Product Code. 10611273 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10611273 1 g Glass bottle 1g
10588843 250 mg Glass bottle 250mg
10473375 500 mg Glass bottle 500mg
10039021 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10611273 Supplier Thermo Scientific Acros Supplier No. 367500010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98014-14-3 | C20H16Cl2N4Ru | 484.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98014-14-3
Molecular Formula C20H16Cl2N4Ru
Molecular Weight (g/mol) 484.35
MDL Number MFCD00150005
InChI Key MGAJEYXQYJBLQL-UHFFFAOYSA-L
Synonym cis-bis 2,2'-bipyridine dichlororuthenium ii hydrate, bis bipyridyl dichlororuthenium hydrate, 2,2'-bipyridine-dichlororuthenium-water 2/1/1, cis-dichlorobis 2,2'-bipyridine ruthenium ii, cis-bis 2,2'-bipyridine dichlororuthenium ii monohydrate, cis-bis 2,2-bipyridine dichlororuthenium ii hydrate, cis-bis-2,2'-bipyridine ruthenium ii chloride,hydrate, cis-bis 2,2 inverted exclamation marka-bipyridine dichlororuthenium ii hydrate
PubChem CID 16211844
SMILES Cl[Ru]Cl.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

Specifications

Color Black to Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Based on Carbon)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble
IUPAC Name dichlororuthenium;2-pyridin-2-ylpyridine;hydrate
Formula Weight 484.35
Percent Purity 97%
Physical Form Powder
Chemical Name or Material cis-Bis(2, 2'-bipyridine)dichlororuthenium(II) hydrate
Show More Show Less
compliance-icons
Product Identifier
  • cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.