missing translation for 'onlineSavingsMsg'
Learn More

cis-Anethole, Mikromol™

Product Code. 18024000
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18024000 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18024000 Supplier Mikromol Supplier No. MM0164.06
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

cis-Anethole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-Anethole
CAS 25679-28-1
Analyte or Component Names cis-Anethole
Shipping Condition Room Temperature
Molecular Formula C10 H12 O
Certifications/Compliance ISO/IEC 17025
Synonym cis-Anethole, Benzene, 1-methoxy-4-(1Z)-1-propen-1-yl-, Anisole, p-propenyl-, (Z)- (8CI), Anisole, p-propenyl-, cis- (7CI), Benzene, 1-methoxy-4-(1-propenyl)-, (Z)-, Benzene, 1-methoxy-4-(1Z)-1-propenyl- (9CI), 1-Methoxy-4-(1Z)-1-propen-1-ylbenzene, (Z)-1-(4-Methoxyphenyl)propene, (Z)-1-Methoxy-4-(prop-1-en-1-yl)benzene, (Z)-Anethole, cis-1-(p-Methoxyphenyl)-1-propene, cis-1-(p-Methoxyphenyl)propene, cis-4-(1-Propenyl)anisole, cis-p-Anethole, cis-p-Methoxy-beta-methylstyrene
InChI Formula InChI=1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3-8H,1-2H3/b4-3-
SMILES COc1ccc(\C=C/C)cc1
IUPAC Name 1-methoxy-4-[(Z)-prop-1-enyl]benzene
Molecular Weight (g/mol) 148.2
Quantity 100 mg
Formula Weight 148.0888 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.