missing translation for 'onlineSavingsMsg'
Learn More

cis-Aconitic acid, tech. 85%

Product Code. 11448626
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448626 5 g 5g
11438626 1 g 1g
11458626 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448626 Supplier Thermo Scientific Alfa Aesar Supplier No. A16010.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 585-84-2 | C6H6O6 | 174.11 g/mol

cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.

Solubility
Soluble in water, alcohol. Slightly soluble in ether.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-84-2
Molecular Formula C6H6O6
Molecular Weight (g/mol) 174.11
MDL Number MFCD00063184
InChI Key GTZCVFVGUGFEME-HNQUOIGGSA-N
Synonym cis-aconitic acid, cis-aconitate, achilleic acid, citridinic acid, equisetic acid, pyrocitric acid, achilleaic acid, citridic acid, cis-aconic acid, aconitic acid
PubChem CID 643757
ChEBI CHEBI:32805
SMILES OC(=O)C\C(=C/C(O)=O)C(O)=O

Specifications

Melting Point 115°C to 120°C
Assay Percent Range tech. 90%
Odor Odorless
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol. Slightly soluble in ether.
IUPAC Name (1E)-prop-1-ene-1,2,3-tricarboxylic acid
Formula Weight 174.11
Percent Purity 90%
Grade Technical
Chemical Name or Material cis-Aconitic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.