missing translation for 'onlineSavingsMsg'
Learn More

cis-9,10-Epoxystearic Acid Methyl Ester, TRC

Product Code. 30382448
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382448 5 g
30369518 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382448 Supplier Toronto Research Chemicals Supplier No. TRCE5898905G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2566-91-8
Molecular Formula C19H36O3
Molecular Weight (g/mol) 312.49
Synonym (2R,3S)-rel-3-Octyl-2-oxiraneoctanoic acid methyl ester, cis- 9,10-Epoxy-octadecanoic acid methyl ester, (2R,3S)-rel-3-Octyl-oxiraneoctanoic acid methyl ester, cis-3-Octyl-xiraneoctanoic acid methyl ester, Methyl 9,10-Epoxyoleate, Methyl cis-9,10-Epoxyoctadecanoate, Methyl cis-9,10-Epoxystearate, Methyl Epoxyoleate
IUPAC Name methyl 8-[(2R,3S)-3-octyloxiran-2-yl]octanoate
SMILES OC[C@H](C)C(C)(C)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C19H36O3/c1-3-4-5-6-8-11-14-17-18(22-17)15-12-9-7-10-13-16-19(20)21-2/h17-18H,3-16H2,1-2H3/t17-,18+/m0/s1
Formula Weight 312.2664
Chemical Name or Material cis-9,10-Epoxystearic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.